missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-8-sulfonic acid, 98%

Product Code. 11316936
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11316936 50 g 50g
11306936 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11316936 Supplier Thermo Scientific Alfa Aesar Supplier No. L06640.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 85-48-3 | C9H7NO3S | 209.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-48-3
Molecular Formula C9H7NO3S
Molecular Weight (g/mol) 209.219
MDL Number MFCD00024037
InChI Key SXVRECLPTCOMIA-UHFFFAOYSA-N
Synonym 8-quinolinesulfonic acid, quinoline-8-sulphonic acid, unii-84y9b81o6x, ccris 6936, 8-sulfoquinoline, cambridge id 5135395, quinoline-8-sulfonic??acid, ksc242e8p, 8-quinolinesulfonic acid;quinoline-8-sulfonic acid
PubChem CID 66561
IUPAC Name quinoline-8-sulfonic acid
SMILES C1=CC2=C(C(=C1)S(=O)(=O)O)N=CC=C2

Specifications

Melting Point >300°C
Quantity 50 g
UN Number UN2585
Beilstein 9501
Formula Weight 209.23
Percent Purity 98%
Chemical Name or Material Quinoline-8-sulfonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.