missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-6-boronic acid, 97%, Thermo Scientific™

Product Code. 15429647
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15429647

Brand: Thermo Scientific Alfa Aesar H52867.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 376581-24-7
Molecular Formula C9H8BNO2
Molecular Weight (g/mol) 172.98
MDL Number MFCD03093100
InChI Key JLOLSBLXNMVKGY-UHFFFAOYSA-N
Synonym quinoline-6-boronic acid, 6-quinolineboronic acid, 6-quinolinylboronic acid, boronic acid, 6-quinolinyl, quinolin-6-yl-6-boronic acid, quinolin-6-yl boronic acid, quinolin-6-ylboronicacidhydrochloride, pubchem1880, 6-quinolylboronic acid, quinoline-6-boronicacid
PubChem CID 4192671
IUPAC Name quinolin-6-ylboronic acid
SMILES OB(O)C1=CC2=CC=CN=C2C=C1

Specifications

Melting Point 106°C to 109°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 172.98
Percent Purity 97%
Chemical Name or Material Quinoline-6-boronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.