missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-6-boronic acid, 97%, Thermo Scientific Chemicals

Product Code. 10658809
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 376581-24-7
Molecular Formula C9H8BNO2
Molecular Weight (g/mol) 172.98
MDL Number MFCD03093100
InChI Key JLOLSBLXNMVKGY-UHFFFAOYSA-N
Synonym quinoline-6-boronic acid, 6-quinolineboronic acid, 6-quinolinylboronic acid, boronic acid, 6-quinolinyl, quinolin-6-yl-6-boronic acid, quinolin-6-yl boronic acid, quinolin-6-ylboronicacidhydrochloride, pubchem1880, 6-quinolylboronic acid, quinoline-6-boronicacid
PubChem CID 4192671
IUPAC Name quinolin-6-ylboronic acid
SMILES OB(O)C1=CC2=CC=CN=C2C=C1

Specifications

Melting Point 144.0°C to 145.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 172.98
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Quinoline-6-boronic acid
compliance-icons
Product Identifier
  • Quinoline-6-boronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'