missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-5-carboxaldehyde, 97+%

Product Code. p-7047698
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11325445

Brand: Thermo Scientific Alfa Aesar L19641.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22934-41-4 | C10H7NO | 157.172 g/mol

Quinoline-5-carboxaldehyde is used as a precursor in the preparation of pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinoline-5-carboxaldehyde is used as a precursor in the preparation of pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22934-41-4
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.172
MDL Number MFCD00805835
InChI Key BNEVFKZLYCGDFG-UHFFFAOYSA-N
Synonym quinoline-5-carboxaldehyde, 5-quinolinecarboxaldehyde, 5-formylquinoline, 5-quinolinecarbaldehyde, acmc-1cmc3, quc
PubChem CID 3543386
IUPAC Name quinoline-5-carbaldehyde
SMILES C1=CC(=C2C=CC=NC2=C1)C=O

Specifications

Melting Point 94°C to 98°C
Quantity 250 mg
Beilstein 113095
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 157.17
Percent Purity ≥97%
Chemical Name or Material Quinoline-5-carboxaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.