missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10632053
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10632053 1 g 1g
10110922 250 mg 250mg
10681236 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10632053 Supplier Thermo Scientific Maybridge Supplier No. CC04601DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 486-74-8
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
InChI Key VQMSRUREDGBWKT-UHFFFAOYSA-N
Synonym 4-quinolinecarboxylic acid, cinchoninic acid, 4-carboxyquinoline, cinchonic acid, usaf d-2, quinoline-4-carboxylicacid, 4-quinolinecarboxylicacid, cinchoninsaure, pubchem5843, chinolin-4-carbonsaeure
PubChem CID 10243
ChEBI CHEBI:18311
IUPAC Name quinoline-4-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CC=N2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 173.17
Percent Purity 97%
Chemical Name or Material Quinoline-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.