missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-4-carboxaldehyde, 97+%

Product Code. 11319046
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319046 25 g 25g
11309046 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319046 Supplier Thermo Scientific Alfa Aesar Supplier No. L20203.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4363-93-3 | C10H7NO | 157.17 g/mol

4-Quinolinecarboxaldehyde(cinchoninaldehyde) was used in the synthesis of lepidylamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Quinolinecarboxaldehyde(cinchoninaldehyde) was used in the synthesis of lepidylamines.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store under dry inert gas. Incompatible with oxidizing agents and air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4363-93-3
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.17
MDL Number MFCD00006781
InChI Key MGCGJBXTNWUHQE-UHFFFAOYSA-N
Synonym 4-quinolinecarboxaldehyde, 4-quinolinecarbaldehyde, cinchoninaldehyde, quinoline-4-carboxaldehyde, 4-formylquinoline, unii-3a0k4yx051, chinolin-4-aldehyd, quinoline-4-aldehyde, 4quinolinecarboxaldehyde
PubChem CID 78072
IUPAC Name quinoline-4-carbaldehyde
SMILES O=CC1=C2C=CC=CC2=NC=C1

Specifications

Melting Point 48°C to 52°C
Boiling Point 131°C to 133°C (5 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 113072
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 157.17
Percent Purity ≥97%
Chemical Name or Material Quinoline-4-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.