missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-3-carboxaldehyde, 98+%

Product Code. 11363267
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11363267 250 mg 250mg
11343267 1 g 1g
11353267 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11363267 Supplier Thermo Scientific Alfa Aesar Supplier No. L13815.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13669-42-6 | C10H7NO | 157.172 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13669-42-6
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.172
MDL Number MFCD00006768
InChI Key RYGIHSLRMNXWCN-UHFFFAOYSA-N
Synonym 3-quinolinecarboxaldehyde, quinoline-3-carboxaldehyde, 3-quinolinecarbaldehyde, 3-formylquinoline, quinoline-3-aldehyde, quinoline-3 carbaldehyde, rarechem ak ml 0129, 3-quinoline carboxaldehyde, quinoline 3-carboxaldehyde, 3-quinoline-carboxaldehyde
PubChem CID 83641
IUPAC Name quinoline-3-carbaldehyde
SMILES C1=CC=C2C(=C1)C=C(C=N2)C=O

Specifications

Melting Point 68°C to 71°C
Quantity 250 mg
Beilstein 113269
Sensitivity Air Sensitive
Formula Weight 157.17
Percent Purity ≥98%
Chemical Name or Material Quinoline-3-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.