missing translation for 'onlineSavingsMsg'
Learn More

Quinine, TRC

Product Code. 30134456
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134456 5 g 5g
30132940 10 g 10g
17934665 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134456 Supplier Toronto Research Chemicals Supplier No. TRCQ6940005G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130-95-0
Molecular Formula C20 H24 N2 O2
Molecular Weight (g/mol) 324.42
Synonym Cinchonan-9-ol, 6'-methoxy-, (8α,9R)-, Quinine (8CI), (8α,9R)-6'-Methoxycinchonan-9-ol, (-)-Quinine, (8S,9R)-Quinine, (R)-(-)-Quinine, (R)-(6-Methoxy-4-quinolyl)[(2S)-5-vinylquinuclidin-2-yl]methanol, 6'-Methoxycinchonidine, Cinkona, Mosgard, NSC 192949, QSM, Qualaquin, Quine 9, Quinex, Quinlex, Quinlup, Rezquin, SW 85833, WR297608, (R)-[(2S,4S,5R)-5-Ethenyl-1-azabicyclo[2.2.2]oct-2-yl]-(6-methoxyquinolin-4-yl)methanol
IUPAC Name (R)-[(2S,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol
SMILES COc1ccc2nccc([C@@H](O)[C@@H]3C[C@@H]4CCN3C[C@@H]4C=C)c2c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C20H24N2O2/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3/t13-,14-,19-,20+/m0/s1
Formula Weight 324.1838
Percent Purity >95%
Chemical Name or Material Quinine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.