missing translation for 'onlineSavingsMsg'
Learn More

Quinine Sulfate Dihydrate, Pure, Fisher Chemical

Product Code. 10774571 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass Jar
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10774571 25 g Glass Jar 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10774571 Supplier Fisher Chemical Supplier No. Q/0136/46
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 6119-70-6
Melting Point 233°C
Color White
pH 5.7 to 6.6
Packaging Glass Jar
Molecular Formula C40H54N4O10S
MDL Number MFCD00150790
Quantity 25 g
InChI Key ZHNFLHYOFXQIOW-OIGVVMIYNA-N
SMILES O.O.OS(O)(=O)=O.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
IUPAC Name (R)-[(2R,4R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;sulfuric acid;tetrahydrate
Molecular Weight (g/mol) 782.95
PubChem CID 134129495
Formula Weight 782.96g/mol
Physical Form Solid
Show More Show Less
compliance-icons
Product Identifier
  • Quinine sulfate dihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.