missing translation for 'onlineSavingsMsg'
Learn More

Quinine monohydrochloride dihydrate, 99%

Product Code. 10458720 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10458720 25 g Glass bottle 25g
10733344 5 g Glass bottle 5g
10658894 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10458720 Supplier Thermo Scientific Acros Supplier No. 163720250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 6119-47-7 | C20H29ClN2O4 | 396.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6119-47-7
Molecular Formula C20H29ClN2O4
Molecular Weight (g/mol) 396.91
MDL Number MFCD00151248
InChI Key MPQKYZPYCSTMEI-VQYSEXPNNA-N
Synonym quinine hcl dihydrate
PubChem CID 124080947
IUPAC Name (R)-[(2S,4S,5S)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;dihydrate;hydrochloride
SMILES O.O.Cl.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C

Specifications

Melting Point 115.0°C to 116.0°C
Color White
Flash Point 122°C
Loss on Drying 6% to 10% (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Fieser 07,311
Merck Index 15, 8177
Specific Rotation Condition − 258.00 (20.00°C c=2,0.1M HCL)
Specific Rotation − 258.00
Solubility Information Solubility in water: 62.5g/L (20 c). Other solubilities: soluble in alcohol,glycerol and chloroform,slightly soluble in ether
Formula Weight 396.92
Percent Purity 99%
Physical Form Crystalline Powder or Silky Needles
Chemical Name or Material Quinine monohydrochloride dihydrate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Quinine monohydrochloride dihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.