missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinine Monohydrochloride Dihydrate, 100.8%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| CAS | 6119-47-7 |
| Molecular Formula | C20H29ClN2O4 |
| MDL Number | MFCD00151248 |
| Quantity | 5 g |
| Synonym | quinine hydrochloride dihydrate, quinine monohydrochloride dihydrate, cinchonan-9-ol, 6'-methoxy-, monohydrochloride, dihydrate, 8a,9r, quinine hcl dihydrate, r-2s,4s,5r-5-ethenyl-1-azabicyclo 2.2.2 octan-2-yl-6-methoxyquinolin-4-yl methanol;dihydrate;hydrochloride, chinini hydrochloridum, quinine, hcl dihydrate, quinine hydrochloride jan:nf, quinine monohydrochloride hydrate |
| InChI Key | MPQKYZPYCSTMEI-VQYSEXPNNA-N |
| SMILES | O.O.Cl.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C |
| IUPAC Name | (R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol dihydrate hydrochloride |
| Molecular Weight (g/mol) | 396.91 |
| PubChem CID | 134690644 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction