missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinine hydrochloride dihydrate, MedChemExpress
Description
Quinine hydrochloride dihydrate (Qualaquin) is an orally active and can be used in anti-malarial studies. Quinine hydrochloride dihydrate is a potassium channel inhibitor that inhibits WT mouse Slo3 (KCa5.1) channel currents evoked by voltage pulses to +100 mV with an IC50 of 169 μM.
Specifications
Specifications
| Chemical Name or Material | Quinine hydrochloride dihydrate |
| Physical Form | Solid |
| CAS | 6119-47-7 |
| Color | White |
| Quantity | 10 g |
| Purity Grade Notes | Research |
| Molecular Formula | C20H29ClN2O4 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : ≥ 100 mg/mL (251.95 mM) ∣H2O : 20 mg/mL (50.39 mM; Need ultrasonic) |
| SMILES | COC1=CC2=C(N=CC=C2[C@H]([C@@]3([H])[N@](C[C@@H]4C=C)CC[C@@]4([H])C3)O)C=C1.Cl.O.O |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?