Learn More
Quinine, anhydrous, 99% (total base), may cont. up to 5% dihydroquinine
Description
Quinine is used in photochemistry as a common fluorescence standard and as a resolving agent for chiral acids. It is also useful for treating falciparum malaria, lupus, arthritis and vivax malaria. It acts as a flavor component in tonic water and bitter lemon. It is utilized as the chiral moiety for the ligands used in sharpless asymmetric dihydroxylation. Furthermore, it catalyzes the kinetic resolution of furanones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Quinine is used in photochemistry as a common fluorescence standard and as a resolving agent for chiral acids. It is also useful for treating falciparum malaria, lupus, arthritis and vivax malaria. It acts as a flavor component in tonic water and bitter lemon. It is utilized as the chiral moiety for the ligands used in sharpless asymmetric dihydroxylation. Furthermore, it catalyzes the kinetic resolution of furanones.
Solubility
Soluble in alcohol, chloroform and diethyl ether. Slightly soluble in water and glycerol.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∼177°C (decomposition) |
| Quantity | 50 g |
| Beilstein | 91867 |
| Sensitivity | Light Sensitive |
| Merck Index | 14,8061 |
| Solubility Information | Soluble in alcohol,chloroform and diethyl ether. Slightly soluble in water and glycerol. |
| Optical Rotation | −166° (c=1 in ethanol) |
| IUPAC Name | (5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl)-[6-(trideuteriomethoxy)quinolin-4-yl]methanol |
| Formula Weight | 324.42 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
- Quinine, anhydrous, 99% (total base), may cont. up to 5% dihydroquinine
- Warning
- Acute toxicity Category 4
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- MIXTURE LIST-Contains : Quinine
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.