missing translation for 'onlineSavingsMsg'
Learn More

Quinine, 95%, MP Biomedicals™

Product Code. 11436172 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
Unit Size:
100g
10g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436172 10 g 10g
11446172 25 g 25g
11456172 50 g 50g
11466172 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11436172 Supplier MP Biomedicals Supplier No. 0210279210
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 130-95-0
Molecular Formula C20H24N2O2
Molecular Weight (g/mol) 324.42
MDL Number MFCD00198096
InChI Key LOUPRKONTZGTKE-ZCXAXGJJNA-N
Synonym quinine, chinin, chinine, --quinine, qualaquin, 6'-methoxycinchonidine, 8s,9r-quinine, chininum, quinine sulfate, quinina
PubChem CID 129316724
IUPAC Name (R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol
SMILES [H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C

Specifications

Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.