missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinidine, TRC
Specifications
Specifications
| Chemical Name or Material | Quinidine |
| CAS | 56-54-2 |
| Molecular Formula | C20 H24 N2 O2 |
| Quantity | 5 g |
| Synonym | Quinidine Sulfate Imp. A (EP), Quinidine Imp. A (EP), Quinine Dihydrochloride Imp. A (EP), Quinine Imp. A (EP), Quinidine, (+)-Quinidine, (8R,9S)-Quinidine, Chinidin, Cinquin, Conchinin, Conchinine, Conquinine, Kinidin, Pitayin, Pitayine, Quindine, beta-Quinidine, beta-Quinine, Quinine Hydrochloride Impurity A, Quinine Impurity A |
| InChI Formula | InChI=1S/C20H24N2O2/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3/t13-,14-,19+,20-/m0/s1 |
| SMILES | COc1ccc2nccc([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)c2c1 |
| IUPAC Name | (S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol |
| Molecular Weight (g/mol) | 324.42 |
| Formula Weight | 324.1838 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Informativa sulla privacy.
Individuate un'opportunità di miglioramento?