missing translation for 'onlineSavingsMsg'
Learn More

Quinidine sulfate dihydrate, 98+%

Product Code. 15443199
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15443199 25 g 25g
15433199 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15443199 Supplier Thermo Scientific Alfa Aesar Supplier No. J60426.14
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A sodium channel blocker, and to a lesser extent also blocks potassium channels | CAS: 6591-63-5 | C40H54N4O10S | 782.95 g/mol

Quinidine sulfate dihydrate, 98+% is used as a A Na+ channel blocker.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinidine sulfate dihydrate, 98+% is used as a A Na+ channel blocker.

Solubility
Soluble in chloroform, water (11 mg/ml at 20°C), alcohol, and ethanol (50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Keep away fom strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6591-63-5
Molecular Formula C40H54N4O10S
Molecular Weight (g/mol) 782.95
MDL Number MFCD00149346
InChI Key ZHNFLHYOFXQIOW-CMZPEELOSA-N
Synonym bis s-6-methoxyquinolin-4-yl 1s,2s,4s,5r-5-vinylquinuclidin-2-yl methanol sulfate dihydrate
PubChem CID 132274356
IUPAC Name (S)-[(2S,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;sulfuric acid;dihydrate
SMILES COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CCN3CC4C=C)O.COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CCN3CC4C=C)O.O.O.OS(=O)(=O)O

Specifications

Melting Point 212°C to 214°C (decomposition)
Color White to Yellow
Odor Odorless
Quantity 25 g
Sensitivity Light sensitive
Merck Index 14,8060
Solubility Information Soluble in chloroform,water (11mg/ml at 20°C),alcohol,and ethanol (50mg/ml).
Formula Weight 782.94
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Quinidine sulfate dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.