missing translation for 'onlineSavingsMsg'
Learn More

Quinidine, MedChemExpress

Product Code. 16468806
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468806 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16468806 Supplier Medchem Express Supplier No. HYB1751
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Quinidine is an antiarrhythmic agent. Quinidine is a potent, orally active, selective cytochrome P450db inhibitor. Quinidine is also a K+ channel blocker with an IC50 of 19.9 μM, and can induce apoptosis. Quinidine can be used for malaria research.

Quinidine is an antiarrhythmic agent. Quinidine is a potent, orally active, selective cytochrome P450db inhibitor. Quinidine is also a K+ channel blocker with an IC50 of 19.9 μM, and can induce apoptosis. Quinidine can be used for malaria research.

Specifications

Chemical Name or Material Quinidine
Physical Form Powder
CAS 56-54-2
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H24N2O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 50 mg/mL (154.12 mM) ∣Ethanol : 14.29 mg/mL (44.05 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 0.67 mg/mL (2.07 mM; Need ultrasonic)
SMILES C=C[C@H]1C[N@](CC[C@H]1C2)[C@@]2([H])[C@@H](O)C3=CC=NC4=CC=C(OC)C=C34
Molecular Weight (g/mol) 324.42
Formula Weight 324.42
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.