Learn More
Thermo Scientific Eutech Quinhydrone ORP solution, 86 mV, ±30 mV @ 25 ºC, 480 mL
Brand:  Thermo Scientific Eutech 01X211215
Quinhydrone ORP solution, 86 mV, ±30 mV @ 25 ºC, 480 mL
Product Code. 11519094