missing translation for 'onlineSavingsMsg'
Learn More

Quinhydrone, Extra Pure, SLR, Fisher Chemical

Product Code. 10070340 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10070340 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10070340 Supplier Fisher Chemical Supplier No. Q/0120/48
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 106-34-3
Melting Point 168°C
Color Green
Packaging Plastic powder jar
Molecular Formula C12H10O4
MDL Number MFCD00010310
Quantity 100 g
Synonym quinhydrone, green hydroquinone, p-benzoquinhydrone, chinhydron, chinhydron czech, unii-p4a66lq3qj, hydroquinone, compd. with p-benzoquinone, p-benzoquinone, compd. with hydroquinone, 2,5-cyclohexadiene-1,4-dione, compd. with 1,4-benzenediol 1:1, p-benzoquinone-hydroquinone compound 1:1
InChI Key BDJXVNRFAQSMAA-UHFFFAOYSA-N
SMILES OC1=CC=C(O)C=C1.O=C1C=CC(=O)C=C1
IUPAC Name benzene-1,4-diol;cyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol) 218.21
PubChem CID 7801
ChEBI CHEBI:26491
Formula Weight 218.21g/mol
Physical Form Powder
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Quinhydrone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.