missing translation for 'onlineSavingsMsg'
Learn More

Quinhydrone, 98.37%, MP Biomedicals™

Product Code. 11470831 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470831 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11470831 Supplier MP Biomedicals Supplier No. 0215648225
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 106-34-3
Color Green
Assay Percent Range 0.9837
Molecular Formula C12H10O4
MDL Number MFCD00010310
Quantity 25 g
Synonym quinhydrone, green hydroquinone, p-benzoquinhydrone, chinhydron, chinhydron czech, unii-p4a66lq3qj, hydroquinone, compd. with p-benzoquinone, p-benzoquinone, compd. with hydroquinone, 2,5-cyclohexadiene-1,4-dione, compd. with 1,4-benzenediol 1:1, p-benzoquinone-hydroquinone compound 1:1
InChI Key BDJXVNRFAQSMAA-UHFFFAOYSA-N
SMILES OC1=CC=C(O)C=C1.O=C1C=CC(=O)C=C1
IUPAC Name benzene-1,4-diol; cyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol) 218.21
PubChem CID 7801
ChEBI CHEBI:26491
Formula Weight 218.2
Physical Form Crystalline Powder
Chemical Name or Material Quinhydrone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.