missing translation for 'onlineSavingsMsg'
Learn More

Quinapril Ethyl Ester Maleic Acid Salt, TRC

Product Code. 30415712
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415712 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415712 Supplier Toronto Research Chemicals Supplier No. TRCQ670015500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Quinapril Ethyl Ester Maleic Acid Salt
CAS 103733-36-4
Molecular Formula C31H38N2O9
Quantity 500 mg
Synonym [3S-[2[R*(R*)],3R*]]-2-[2-[[1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic Acid Ethyl Ester (2Z)-2-Butenedioate
InChI Formula InChI=1S/C27H34N2O5.C4H4O4/c1-4-33-26(31)23(16-15-20-11-7-6-8-12-20)28-19(3)25(30)29-18-22-14-10-9-13-21(22)17-24(29)27(32)34-5-2;5-3(6)1-2-4(7)8/h6-14,19,23-24,28H,4-5,15-18H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t19-,23-,24-;/m0./s1
SMILES CCOC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N2Cc3ccccc3C[C@H]2C(=O)OCC.OC(=O)\C=C/C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;ethyl (3S)-2-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-3,4-dihydro-1H-isoquinoline-3-carboxylate
Molecular Weight (g/mol) 582.64
Formula Weight 582.2577
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.