missing translation for 'onlineSavingsMsg'
Learn More

Quinalphos free hydroxy, Dr. Ehrenstorfer

Product Code. 30311703
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30311703

Brand: Dr. Ehrenstorfer DREC16700100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Quinalphos free hydroxy
CAS 1196-57-2
Analyte or Component Names Quinalphos free hydroxy
Shipping Condition Room Temperature
Molecular Formula C8 H6 N2 O
Certifications/Compliance ISO/IEC 17025
Synonym 2-Quinoxalinol (7CI), 1,2-Dihydroquinoxalin-2-one, 2-Hydroxyquinoxaline, 2-Quinoxalinone, 2-Quinoxalone, 3-Quinoxalinone, NSC 13154, Quinoxaline-2-ol
InChI Formula InChI=1S/C8H6N2O/c11-8-5-9-6-3-1-2-4-7(6)10-8/h1-5H,(H,10,11)
Solution Type Neat
SMILES Oc1cnc2ccccc2n1
IUPAC Name quinoxalin-2-ol
Industry Type Environmental Testing
Molecular Weight (g/mol) 146.15
Quantity 100 mg
Formula Weight 146.048 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.