missing translation for 'onlineSavingsMsg'
Learn More

Quinaldoyl chloride, 95%

Product Code. p-4228991 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11996501

Brand: Thermo Scientific Acros 432320010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 50342-01-3 | C10H6ClNO | 191.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50342-01-3
Molecular Formula C10H6ClNO
Molecular Weight (g/mol) 191.61
MDL Number MFCD02683554
InChI Key WFVMVMAUXYOQSW-UHFFFAOYSA-N
Synonym quinaldoyl chloride, 2-quinolinecarbonyl chloride, quinaldyl chloride 97, quinaldyl chloride, chloroformylquinoline, 2-quinolinecarbonyl chloride 9ci, quinoline-2-carboxylic acid chloride
PubChem CID 2760547
IUPAC Name quinoline-2-carbonyl chloride
SMILES ClC(=O)C1=NC2=CC=CC=C2C=C1

Specifications

Melting Point 94°C to 97°C
Color Orange to Brown
Physical Form Crystalline Powder
Quantity 1 g
Assay Percent Range 94% min. (GC)
Formula Weight 191.62
Percent Purity 95%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Quinaldoyl chloride
compliance-icons
Product Identifier
  • Quinaldoyl chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.