missing translation for 'onlineSavingsMsg'
Learn More

Quinagolide hydrochloride, MedChemExpress

Product Code. 16406306
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16406306

Brand: Medchem Express HY13736A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Quinagolide hydrochloride is a selective dopamine D2 receptor agonist, also is a prolactin inhibitor.

Quinagolide hydrochloride is a selective dopamine D2 receptor agonist, also is a prolactin inhibitor.

Specifications

Chemical Name or Material Quinagolide hydrochloride
CAS 94424-50-7
Color White
Purity Grade Notes Research
Molecular Formula C20H34ClN3O3S
Quantity 100 mg
Synonym CV205-502 hydrochloride
Solubility Information DMSO : 3.85 mg/mL (8.91 mM; Need ultrasonic)
SMILES OC1=C2C(C[C@](N(CCC)C[C@@H](NS(N(CC)CC)(=O)=O)C3)([H])[C@@]3([H])C2)=CC=C1.Cl
Molecular Weight (g/mol) 432.02
Formula Weight 432.02
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.