missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Quin C1
GREENER_CHOICE

Product Code. p-7110957
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18792459

Brand: Tocris Bioscience™ 5583/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective FPR2 agonist

Quin C1 is a potent and selective FPR2 agonist (EC50 = 15 nM). Induces Ca2+ mobilization in FPR2 but not FPR1-transfected mast cells. Induces neutrophil chemotaxis and degranulation in vitro. Reduces neutrophil and lymphocyte counts in bronchoalveolar lavage fluid in a mouse lung injury model. Anti-inflammatory.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Butoxy-N-[1,4-dihydro-2-(4-methoxyphenyl)-4-oxo-3(2H)-quinazolinyl]benzamide
CAS 786706-21-6
Quantity 10 mg
Target Formyl Peptide Receptor Agonists
Molecular Formula C26H27N3O4
Formula Weight 445.51
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.