missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Transport Case for Buerkle™ QuickPicker

Product Code. 10613062 Shop All Bürkle Products
Change view
Click to view available options
For Use With (Equipment):
For QuickPicker
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10613062 For QuickPicker Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10613062 Supplier Buerkle™ Supplier No. 36003000
  • ONLINE EXCLUSIVE PRICE
    £31.90
    £31.35 / Pack of 1
    Save £0.55
    1% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Buerkle™ QuickPicker Transport Case is the solution to transport the Buerkle™ QuickPicker safely.

  • Case for safe transport
  • For use with the Buerkle™ QuickPicker
TRUSTED_SUSTAINABILITY

Specifications

Product Type Transport Case
Color Black, Red
For Use With (Equipment) For QuickPicker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.