Learn More
Quickfit™ Virgeux Fractionating Columns
Click to view available options
Standard Taper Joint:
19/26
24/29
Unit Size:
Each
Description
- Easy to use column
- Vigreux type with combination of horizontal and angled indentations
- Horizontal indentations ensure intimate vapor and liquid contact
- Angled indentations redistribute liquid from walls to center of column
Specifications
Specifications
| Includes | Side arm and screw threads |
| Length (Metric) | 178 mm |
| Type | Fractionating Column |
| Standard Taper Joint | 19/26 |