missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Unbadged Joints Cones
Free from any ceramic marking
Brand: Quickfit CNB45UB
Code : A0
Additional Details : Weight : 0.55000kg
Description
Free from any ceramic marking
- Avoids problem of badge charring at high temperatures which can give an untidy appearance
- Manufactured to accuracy which surpasses requirements of ISO 383
- Pyrex™ borosilicate glass for resistance to chemical corrosion and thermal shock
- Cones manufactured with shank length of 100mm
Specifications
Joint Cone | |
45/40 | |
2.30mm |