Learn More
Quickfit™ Two-Neck Round-Bottom Glass Flask
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Two-Neck Round-Bottom Glass Flask has high resistance to thermal shock and chemical attack.
Brand:  Quickfit FR2L/5S/2A
Product Code. 11371945
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Fitted with two Quickfit™ ground glass sockets - One vertical centre neck - One angled side neck
  • Side neck useful for introducing gas inlet tubes and thermometers etc. , into the flask
  • Manufactured from borosilicate glass for highest resistance to thermal shock and chemical attack