Learn More
Quickfit™ Straight Tubing Adapter
Add detachable tubing to a glass vescle with a glass tubing adapter. QuickFit™ straight tubing adapters come in different sizes to fit containers or varying sizes.
Brand: Quickfit MF13/2/SC
31.39 GBP valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Code : 01
Additional Details : Weight : 0.02632kg
Description
- With straight connection for the attachment of rubber hose etc
- Detachable plastic screwthread connectors allow flexible tubing to be fitted easily and safely
Specifications
Flexible tubing, Rubber hose | |
Borosilicate Glass | |
19/26 |
Detachable Plastic screw thread connector | |
Straight tubing adapter |