Learn More
SVL™ Socket/Screw Joint Adapter
Product Code. 15557740
Change view
Click to view available options
Standard Taper Joint No.:
15
22
30
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No. unitSize
15557740 22 Pack of 10
15547740 15 Pack of 10
15567740 30 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15557740 Supplier SVL Supplier No. 67003
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Facilitates connection between SVL™ and Quickfit™ glassware.
  • Features an SVL™ screwthread and Quickfit™ ground glass socket
  • Thread accepts SVL™ screw caps and couplings. Socket fits Quickfit™ cones
  • Manufactured from temperature and chemical resistant borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) SVL™ and Quickfit™ Glassware
Material Borosilicate Glass
Product Type Socket/screw thread ground joint adapter
Standard Taper Joint No. 22