Learn More
Quickfit™ Tubing Adapter with Side-arm
Quickfit™ Tubing Adapter with Side-arm for attachment of rubber hose
Brand:  Quickfit MF17/2/SC
Code : NEW
Additional Details : Weight : 0.03000kg
 View more versions of this product
Product Code. 15587930
  • £144.00 / Pack of 5
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
Description
Description
Quickfit™ Tubing Adapter with Side-arm for attachment of rubber hose
  • With sidearm for the attachment of rubber hose etc
  • Detachable plastic screwthread connector allows flexible tubing to be fitted easily and safely
Specifications
Specifications
19/26
Borosilicate Glass
14/23
Detachable Plastic screw thread connector
Side-arm tubing adapter