Learn More
Quickfit™ Tubing Adapter with Side-arm
Quickfit™ Tubing Adapter with Side-arm for attachment of rubber hose
Brand:  Quickfit MF18/44/SC
38.79 GBP valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Code : 01
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 12455316
  • £91.30 / Each
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
Description
Description
Quickfit™ Tubing Adapter with Side-arm for attachment of rubber hose
  • With sidearm for the attachment of rubber hose etc
  • Detachable plastic screwthread connector allows flexible tubing to be fitted easily and safely
Specifications
Specifications
29/32
Borosilicate Glass
29/32
Detachable Plastic screw thread connector
Side-arm tubing adapter