Learn More
Quickfit™ Tubing Adapter with Side-arm
Product Code. 12028371
Change view
Click to view available options
Cone Size:
14/23
19/26
24/29
29/32
34/35
NS 24/29
Standard Taper Joint No.:
14/23
19/26
24/29
29/32
NS 24/29
Unit Size:
Each
Pack of 5
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cone Size Standard Taper Joint No. unitSize
12028371 14/23 14/23 Each
10440485 19/26 19/26 Each
12048371 24/29 19/26 Each
12058371 24/29 24/29 Each
12455316 29/32 29/32 Each
12465316 34/35 19/26 Each
15507940 29/32 19/26 Pack of 5
15587930 19/26 14/23 Pack of 5
15597930 NS 24/29 NS 24/29 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 12028371 Supplier Quickfit Supplier No. MF17/1/SC
Only null left
Add to basket
Add to basket
Quickfit™ Tubing Adapter with Side-arm for attachment of rubber hose
  • With sidearm for the attachment of rubber hose etc
  • Detachable plastic screwthread connector allows flexible tubing to be fitted easily and safely
TRUSTED_SUSTAINABILITY
Specifications
Cone Size 14/23
Includes Detachable Plastic screw thread connector
Material Borosilicate Glass
Product Type Side-arm tubing adapter
Standard Taper Joint No. 14/23