Learn More
Quickfit™ Sloping Splash Head Adapter
Used in distillation assemblies.
Brand: Quickfit SH7/0/44
Code : NEW
Additional Details : Weight : 0.14000kg
Description
- Used in distillation assemblies to prevent raw liquid carrying over from flask to condenser
Specifications
Adapter | |
Used in Distillation Assemblies to Prevent Raw liquid “Carrying over” From Flask to Condenser | |
1 Pack | |
29/32 (Condenser), 29/32 (flask) |