Learn More
PYREX™ Quickfit™ Steam Distillation Splash Head
Product Code. 15587960
Change view
Click to view available options
Taper:
Cone: 19/26, Socket: 29/32
Cone: 29/32, Socket: 29/32
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Taper Unit Size
15587960 Cone: 19/26, Socket: 29/32 Pack of 5
13437656 Cone: 29/32, Socket: 29/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15587960 Supplier Quickfit Supplier No. SH16/19
  • ONLINE EXCLUSIVE PRICE
    £641.55
    £625.74 / Pack of 5
    Save £15.81
    2% Off
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
Distillation head with steam connection
  • For use with rotary evaporators
  • Prevents possible contamination of the condenser from 'bumping' solvents
  • Features a coarse porosity 1 sinter to reduce carry over
  • Splash heads feature a 29/32 socket
Specifications
Description Anti-splash with sinter
Material Borosilicate Glass
Type Splash head adapter
For Use With (Application) Prevents possible contamination of the condenser from 'bumping' solvents
For Use With (Equipment) Rotary evaporators
Includes Coarse, porosity 1 sinter
Quantity 1 Each
Taper Cone: 19/26, Socket: 29/32
Volume (Metric) 90 mL