Learn More
Quickfit™ PTFE Stirrer Gland
Product Code. 12013314
Change view
Click to view available options
Includes:
Integral drive pulley, Brass cone
PTFE cone
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
12013314 PTFE cone Each
12445416 Integral drive pulley, Brass cone Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12013314 Supplier Quickfit Supplier No. ST3/2F
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Create a tight seal with PTFE stirrer glands. Quickfit™ PTFE Stirrer Glands provide a seal to a stirrer shaft by the adjustable compression of rubber 'O' rings and felt washers impregnated with lubricant.
Create a tight seal with PTFE stirrer glands. QuickfitTM PTFE Stirrer Glands provide a seal to a stirrer shaft by the adjustable compression of rubber 'O' rings and felt washers impregnated with lubricant.
  • 'Stuffing box' glands available with either brass or PTFE cones
  • Glands ST4/2 and ST4/2F have integral drive pulleys
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 35 mm Pully
For Use With (Equipment) Fits 6mm stirrer shaft
Includes PTFE cone
Standard Taper Joint 19/26
Type Stirrer Gland
Material PTFE