Learn More
Quickfit™ PTFE Clamp
Product Code. p-8002686
Click to view available options
Size:
14/23
29/32
Unit Size:
Pack of 5
Product Code. 11941528
Brand: Quickfit GC14
Only null left
Add to basket
Add to basket
For retaining glassware having Quickfit™ conical joints
  • Used for retaining glassware having Quickfit™ conical joints
  • Manufactured from PTFE with encapsulated steel spring to provide firm grip
  • Clips are numbered with joint size for easy identification
  • Can be used at temperatures up to 250°C
TRUSTED_SUSTAINABILITY
Specifications
Type Joint Clamp
Material PTFE
Includes Encapsulated Steel spring
For Use With (Equipment) Conical Joints
Size 14/23
Temperature (Metric) Resistant 250°C