Learn More
Quickfit™ PTFE Clamp
Description
- Used for retaining glassware having Quickfit™ conical joints
- Manufactured from PTFE with encapsulated steel spring to provide firm grip
- Clips are numbered with joint size for easy identification
- Can be used at temperatures up to 250°C
Specifications
Specifications
| Type | Joint Clamp |
| Material | PTFE |
| Includes | Encapsulated Steel spring |
| For Use With (Equipment) | Conical Joints |
| Size | 29/32 |
| Temperature (Metric) Resistant | 250°C |