missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ PTFE Blade

Use these PTFE blades for maximum resistance to chemical attack. Quickfit™ PTFE Blades can be pivoted on the glass shaft to allow passage through small apertures e.g. necks and sockets of Quickfit™ flasks.

Brand:  Quickfit ST7/3L

Code : 01

Additional Details : Weight : 1.18000kg

 View more versions of this product

Product Code. 12939413

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Use these PTFE blades for maximum resistance to chemical attack. QuickfitTM PTFE Blades can be pivoted on the glass shaft to allow passage through small apertures e.g. necks and sockets of QuickfitTM flasks.

  • Blades are detachable for cleaning and replacement purposes
  • Useful for sweeping close to the vessel walls as they will not cause scratching
  • For use with glass stirrer shafts
Specifications

Specifications

11 mm
For sweeping close to the vessel walls as they will not cause scratching
250°C
76mm (Blade)
Glass stirrer shaft
750mm
Stirrer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers