Learn More
Scilabware Quickfit™ Multiple Glass Joint Adapter with Two Parallel Necks
Connect two apparatus together leak-tight. The QuickFit™ multiple glass joint adapter with two parallel necks is the perfect choice.
Brand: Scilabware MA1/44
Code : A0
Additional Details : Weight : 0.11000kg
Description
Connect two apparatus together leak-tight. The QuickFit™ multiple glass joint adapter with two parallel necks is the perfect choice.
•With two parallel necksSpecifications
29/32 | |
Borosilicate Glass | |
Multiple neck adapter | |
29/32 |