Learn More
Quickfit™ Metal Clamps for Spherical Joints
Description
- Suitable for high temperature work
- Spring loaded to ensure balanced positive pressure to both the ball and socket joint surface
- Available in a variety of sizes
Specifications
Specifications
| Quantity | 5/Pk. |
| Type | Spherical Joint Clamp |
| Material | Nickel-plated Metal |
| For Use With (Equipment) | Spherical joint size S19 |