Learn More
Quickfit™ Hexagonal Hollow-blown Glass Stopper
Description
Ideally suited for use with volumetric flasks.
- Lower closed end of stopper features 'drip-tip' to ensure adhered liquid is returned to flask
- Hexagonal head prevents stopper from rolling and provides good grip
- Flat head allows stopper to be stood on end, thus preventing contamination of ground surface
- Hollow-blown to reduce weight
Specifications
Specifications
| For Use With (Equipment) | Volumetric flasks |
| Standard Taper Joint | 10/19 |
| Material | Glass |
| Product Type | Hollow Stopper |