Learn More
Quickfit™ Glass Funnel Cap for Markham Still Nitrogen Determination Distillation Unit
Click to view available options
Material:
Glass
Unit Size:
Each
Specifications
Specifications
| For Use With (Equipment) | Nitrogen determination distillation unit |
| Standard Taper Joint | 10/19 |
| Material | Glass |
| Product Type | Funnel Cap |