Learn More
Quickfit™ Expansion Socket Adapter
Made of borosilicate glass. Connects large sized ground glass joints with smaller sized ones. With the QuickFit™ expansion socket adaptor.
Brand: Quickfit XA64
Code : Z4
Additional Details : Weight : 0.08000kg
Description
Connect large sized ground glass joints with smaller sized ones with the QuickFit™ expansion socket adaptor
- Useful for the connection of dissimilar sized ground glass joints
- Large sized socket at the top and small sized cone at the base
Specifications
29/32 | |
Borosilicate Glass | |
40/38 |
Useful for the connection of dissimilar sized ground Glass joints | |
Expansion adapter |