Learn More
Quickfit™ Chromatography Column with Ground Cone
Product Code. p-6806152
Click to view available options
Length (Metric):
310 mm
515 mm
Unit Size:
Pack of 5
Product Code. 15537770
Brand: Quickfit CR32/20
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from borosilicate glass. Quickfit™ Chromatography Column with Ground Cone includes a 0 porosity chemically resistant glass sintered disc. Upper socket can be connected to a solvent reservoir. Lower cone can be connected to stopcock adapter CR3T/23 to control eluent flow.
  • With glass sintered disc - porosity 0 to support column packing
  • Upper socket can be connected to a solvent reservoir
  • Lower cone can be connected to stopcock adapter CR3T/23 to control eluent flow
  • Manufactured from borosilicate glass
  • TRUSTED_SUSTAINABILITY
Specifications
Description Chromatography Column
Quantity 1 Pack
Diameter (Metric) Inner 18 mm
Length (Metric) 310 mm
Material Pyrex™ Borosilicate Glass
Connector Types 19/26 Cone, 24/29 Socket
Includes Glass sintered disc