Learn More
Quickfit™ Borosilicate Glass Vigreux Fractionating Column
Brand:  Quickfit FC3/23
Product Code. 12051652
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Popular, easy to use column
  • Vigreux type with combination of horizontal and angled indentations
  • Horizontal indentations ensure intimate vapour and liquid contact
  • Angled indentations redistribute liquid from walls to centre of column