Learn More
Quickfit™ Borosilicate Glass Dufton Fractionating Column
Product Code. p-8002933
Change view
Click to view available options
Length (Metric):
270mm
290mm
300mm
425mm
Unit Size:
Each
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
12001652 270mm Each
12408906 425mm Each
15507820 290mm Pack of 5
15517820 300mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12001652 Supplier Quickfit™ Supplier No. FC1/12
Efficient fractionating column designed with an an accurately fitted Dufton glass spiral. Quickfit™ Borosilicate Glass Dufton Fractionating Column can be disassembled for cleaning.
  • Glass column with accurately fitted Dufton glass spiral
  • Good efficiency - liquid spiralling down column meets ascending vapour
  • Spiral can be removed for cleaning
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 270mm
Type Dufton Fractionating Column
Material Borosilicate Glass
Certifications/Compliance ISO 3585