Learn More
Quickfit™ Borosilicate Glass Double Coil Condenser
Brand:  Quickfit C13/55
Product Code. 10130920
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Features a double coil design with a common water inlet
  • Suitable for pilot plant work
  • With two independent cooling coils
  • Each can be fed from separate water supplies for maximum efficiency