Learn More
Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone
Circulates coolant through a coil with a very large cooling surface. Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone is ideal for use in the upright reflux mode where counter-current cooling is possible. Supplied with detachable plastic connectors for the attachment of rubber hose.
Brand: Quickfit CX3/08/SC
Code : NEW
Additional Details : Weight : 0.30000kg
Description
Circulates coolant through a coil with a very large cooling surface. Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone is ideal for use in the upright reflux mode where counter-current cooling is possible. Supplied with detachable plastic connectors for the attachment of rubber hose.
- Coolant is circulated through a coil having a very large cooling surface
- Ideal for use in the upright reflux mode where counter-current cooling is possible
- Supplied with detachable plastic connectors for the attachment of rubber hose
Specifications
50/42 | |
Condenser Coil | |
Borosilicate Glass |
Ideal for use in the upright reflux mode where counter-current cooling is possible | |
348 mm (Overall) 155 mm (Effective) |